Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079669.2 | NP_001073137.1 | 11 | 1586 | ||
NM_001286453.1 | NP_001273382.1 | 9 | 1253 | |||
NM_032813.3 | NP_116202.2 | 12 | 1796 | |||
XM_011521121.2 | XP_011519423.1 | 13 | 1948 | |||
XM_011521122.2 | XP_011519424.1 | 13 | 1864 | |||
XM_011521123.2 | XP_011519425.1 | 13 | 2025 | |||
XM_011521124.2 | XP_011519426.1 | 12 | 1653 | |||
XM_011521125.1 | XP_011519427.1 | 13 | 1857 | |||
XM_017020796.1 | XP_016876285.1 | 12 | 1851 | |||
XM_017020797.1 | XP_016876286.1 | 11 | 1590 | |||
XM_017020798.1 | XP_016876287.1 | 11 | 1686 | |||
XM_017020799.1 | XP_016876288.1 | 10 | 1371 | |||
GenBank mRNA | AF339792.1 | - | 9 | 1168 | ||
AK027530.1 | - | 6 | 679 | |||
AK056409.1 | - | 12 | 1655 | |||
AK092653.1 | - | 4 | 1669 | |||
AK290345.1 | - | 11 | 1446 | |||
AK299859.1 | - | 9 | 1092 | |||
BC018707.1 | - | 6 | 757 | 4841071 | ||
BG830268.1 | - | 4 | 472 | |||
BX647956.1 | - | 11 | 1477 | |||
BX648008.1 | - | 11 | 1430 |