Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C9ORF3 SIRNA Assays ›
Gene Symbol:
C9orf3
Gene Name:
chromosome 9 open reading frame 3
Gene Aliases:
AOPEP, AP-O, APO, C90RF3, ONPEP
Chromosome Location:
Chr.9: 94726604 - 95148264 on Build GRCh38
UniGene ID:
Hs.434253
Species:
Human
siRNA ID
HSS131440
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C9orf3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001193329.1
NP_001180258.1
7, 8
1909
NM_032823.5
NP_116212.3
6, 7
1695
XM_005252284.2
XP_005252341.1
6, 7
1760
XM_006717306.1
XP_006717369.1
8, 9
2057
XM_011519119.1
XP_011517421.1
8, 9
2057
XM_011519120.2
XP_011517422.1
8, 9
2057
XM_011519121.2
XP_011517423.1
8, 9
2057
XM_011519122.2
XP_011517424.1
8, 9
2057
XM_011519123.2
XP_011517425.1
8, 9
2057
XM_011519127.1
XP_011517429.1
8, 9
2057
XM_011519132.1
XP_011517434.1
5, 6
783
XM_017015229.1
XP_016870718.1
8, 9
2100
XM_017015230.1
XP_016870719.1
8, 9
2057
XM_017015231.1
XP_016870720.1
6, 7
1760
XM_017015232.1
XP_016870721.1
8, 9
2057
XM_017015233.1
XP_016870722.1
8, 9
2087
XM_017015234.1
XP_016870723.1
7, 8
1890
XM_017015235.1
XP_016870724.1
7, 8
1890
XM_017015236.1
XP_016870725.1
5, 6
1593
XM_017015237.1
XP_016870726.1
5, 6
1593
GenBank mRNA
AJ560639.1
-
7, 8
1774
AK027581.1
-
5, 6
946
BC020194.1
-
7, 8
1120
5020674
BC029616.1
-
6, 7
1652
3877098
BX648970.1
-
6, 7
995
Back To Top
More Information
Panther Classification:
Molecular Function -
protease
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
peptide catabolic process
aminopeptidase activity
metallopeptidase activity
zinc ion binding
peptide binding
metalloaminopeptidase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0