Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032832.5 | NP_116221.3 | 7 | 1807 | ||
XM_011536194.2 | XP_011534496.1 | 6 | 1709 | |||
GenBank mRNA | AK027641.1 | - | 7 | 1489 | ||
AK299599.1 | - | 7 | 1050 | |||
BC043141.2 | - | 7 | 1808 | 5286304 |