Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032268.4 | NP_115644.1 | 1 | 949 | ||
XM_011523392.1 | XP_011521694.1 | 1 | 923 | |||
XM_017023793.1 | XP_016879282.1 | 1 | 923 | |||
XM_017023794.1 | XP_016879283.1 | 1 | 923 | |||
GenBank mRNA | AF378524.1 | - | 1 | 621 | ||
AK294434.1 | - | 1 | 621 | |||
AL136903.1 | - | 1 | 614 | |||
AL834440.1 | - | 1 | 273 | |||
BC007235.1 | - | 1 | 621 | 2823677 |