Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281504.1 | NP_001268433.1 | 6, 7 | 1228 | ||
NM_032885.5 | NP_116274.3 | 6, 7 | 1154 | |||
NR_104024.1 | 6, 7 | 1110 | ||||
NR_104025.1 | 5, 6 | 877 | ||||
XM_006722927.1 | XP_006722990.1 | 4, 5 | 809 | |||
GenBank mRNA | AJ312332.1 | - | 6, 7 | 970 | ||
AK056210.1 | - | 6, 7 | 1129 | |||
AK223383.1 | - | 6, 7 | 1079 | |||
AK294676.1 | - | 6, 7 | 1228 | |||
AK314712.1 | - | 6, 7 | 1089 | |||
BC007639.1 | - | 5, 6 | 800 | |||
BC068992.1 | - | 6, 7 | 1130 | 6578102 |