Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033087.3 | NP_149078.1 | 2 | 842 | ||
NR_024532.1 | 3 | 1001 | ||||
GenBank mRNA | AB161356.1 | - | 2 | 799 | ||
AK027417.1 | - | 2 | 794 | |||
AK027814.1 | - | 1 | 1594 | |||
AK074704.1 | - | 2 | 794 | |||
AK074988.1 | - | 3 | 979 | |||
AK075172.1 | - | 2 | 818 | |||
AK075189.1 | - | 1 | 62 | |||
AY358697.1 | - | 2 | 586 | |||
BC017876.1 | - | 2 | 792 | 4698763 |