Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271999.1 | NP_001258928.1 | 21 | 2673 | ||
NM_001272000.1 | NP_001258929.1 | 21 | 2673 | |||
NM_001272001.1 | NP_001258930.1 | 21 | 2673 | |||
NM_033407.3 | NP_212132.2 | 21 | 2673 | |||
XM_005271292.2 | XP_005271349.1 | 21 | 2663 | |||
XM_011542326.2 | XP_011540628.1 | 21 | 2663 | |||
XM_011542327.2 | XP_011540629.1 | 21 | 2663 | |||
XM_011542328.2 | XP_011540630.1 | 21 | 2663 | |||
XM_011542330.2 | XP_011540632.1 | 21 | 2663 | |||
XM_017002639.1 | XP_016858128.1 | 21 | 2663 | |||
XM_017002640.1 | XP_016858129.1 | 21 | 2663 | |||
GenBank mRNA | AB051558.1 | - | 1 | 83 | ||
AK125049.1 | - | 7 | 818 | |||
DQ118679.1 | - | 18 | 2245 | |||
DQ118680.1 | - | 18 | 2245 | |||
DQ309763.1 | - | 18 | 2245 | |||
DQ341187.1 | - | 21 | 2580 |