Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC16B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033127.2 | NP_149118.2 | 14 | 1833 | ||
GenBank mRNA | AB063357.1 | - | 14 | 1833 | ||
AK093939.1 | - | 6 | 627 | |||
AK098627.1 | - | 3 | 453 | |||
AK314585.1 | - | 13 | 1837 | |||
BC009106.1 | - | 14 | 1828 | 3448742 | ||
EF125213.1 | - | 13 | 1743 |