Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR3DX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026716.2 | 9 | 1392 | |||
NR_104095.1 | 8 | 1072 | ||||
NR_104096.1 | 8 | 1054 | ||||
NR_104097.1 | 8 | 1165 | ||||
NR_136268.1 | 9 | 1392 | ||||
GenBank mRNA | AK024467.1 | - | 4 | 2212 | ||
BC033195.1 | - | 4 | 427 | |||
BC171827.1 | - | 8 | 1072 | |||
BC171832.1 | - | 8 | 1054 | |||
BC171834.1 | - | 8 | 1326 | |||
BC171838.1 | - | 8 | 1165 | |||
BC171839.1 | - | 7 | 1267 | |||
DQ224421.1 | - | 9 | 1293 |