Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHLDB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134437.1 | NP_001127909.1 | 10 | 2778 | ||
NM_001134438.1 | NP_001127910.1 | 10 | 2965 | |||
NM_001134439.1 | NP_001127911.1 | 10 | 2726 | |||
NM_145753.2 | NP_665696.1 | 9 | 2836 | |||
GenBank mRNA | AB209903.1 | - | 10 | 2607 | ||
AF506820.1 | - | 8 | 2540 | |||
AJ496194.1 | - | 9 | 2580 | |||
AK296841.1 | - | 10 | 2778 | |||
AK315050.1 | - | 10 | 2876 | |||
BC069194.1 | - | 9 | 2725 | |||
BC142678.1 | - | 9 | 2572 | 40148225 | ||
BC150210.1 | - | 9 | 2572 | 40148898 | ||
CR749654.1 | - | 10 | 2761 |