Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033271.2 | NP_150374.2 | 5 | 953 | ||
XM_005268188.3 | XP_005268245.1 | 4 | 1121 | |||
GenBank mRNA | AF353674.1 | - | 4 | 817 | ||
BC042525.1 | - | 5 | 923 | 4824525 |