Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142301.1 | NP_001135773.1 | 17, 18 | 1685 | ||
NM_153704.5 | NP_714915.3 | 16, 17 | 1754 | |||
NR_024522.1 | 16, 17 | 1754 | ||||
XM_006716686.3 | XP_006716749.1 | 15, 16 | 1592 | |||
XM_011517363.2 | XP_011515665.1 | 7, 8 | 844 | |||
GenBank mRNA | AK026742.1 | - | 11, 12 | 1060 | ||
AK092244.1 | - | 17, 18 | 1642 | |||
AK094935.1 | - | 16, 17 | 1678 | |||
AK297676.1 | - | 1, 2 | 407 | |||
AK308735.1 | - | 7, 8 | 821 | |||
BC032835.2 | - | 16, 17 | 1672 | 4825770 | ||
BC054338.1 | - | 16, 17 | 1681 | |||
BX648768.1 | - | 16, 17 | 1698 | |||
KU178827.1 | - | 16, 17 | 1653 | |||
KU178828.1 | - | 15, 16 | 1499 |