Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033200.2 | NP_149977.2 | 13, 14 | 1837 | ||
XM_005261955.3 | XP_005262012.1 | 13, 14 | 1919 | |||
XM_006724426.3 | XP_006724489.1 | 12, 13 | 2283 | |||
XM_006724427.3 | XP_006724490.1 | 13, 14 | 1834 | |||
XM_017029077.1 | XP_016884566.1 | 12, 13 | 1857 | |||
GenBank mRNA | AK130818.1 | - | 6, 7 | 648 | ||
BC002942.2 | - | 13, 14 | 1811 | 3954961 | ||
BC014652.1 | - | 13, 14 | 1900 | 4562676 | ||
BC021143.1 | - | 13, 14 | 1811 | 4872642 | ||
BX412067.2 | - | 1, 2 | 753 | |||
CR456605.1 | - | 13, 14 | 1821 |