Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195144.1 | NP_001182073.1 | 11 | 1325 | ||
XM_005246945.1 | XP_005247002.1 | 11 | 1315 | |||
XM_005246946.1 | XP_005247003.1 | 11 | 1315 | |||
XM_005246947.2 | XP_005247004.1 | 11 | 1315 | |||
XM_005246948.2 | XP_005247005.1 | 11 | 1315 | |||
XM_006712838.3 | XP_006712901.1 | 11 | 1315 | |||
XM_011512148.2 | XP_011510450.1 | 11 | 1315 | |||
XM_017005252.1 | XP_016860741.1 | 11 | 1315 | |||
GenBank mRNA | AK097086.1 | - | 11 | 1134 | ||
AK301209.1 | - | 10 | 1246 | |||
AL133087.1 | - | 5 | 524 | |||
BC016985.2 | - | 11 | 1299 | 4395011 |