Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASL10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033315.3 | NP_201572.1 | 2, 3 | 570 | ||
XM_017025300.1 | XP_016880789.1 | 2, 3 | 2137 | |||
XM_017025301.1 | XP_016880790.1 | 1, 2 | 1204 | |||
GenBank mRNA | AB047296.1 | - | 1, 2 | 322 | ||
AK122652.1 | - | 2, 3 | 570 | |||
BC041133.1 | - | 2, 3 | 491 | 5762715 |