Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153369.2 | NP_699200.2 | 4 | 1573 | ||
XM_017011483.1 | XP_016866972.1 | 3 | 1456 | |||
GenBank mRNA | AB067506.1 | - | 1 | 808 | ||
AK093136.1 | - | 4 | 1532 | |||
AK292735.1 | - | 4 | 1502 | |||
AK315016.1 | - | 4 | 1497 | |||
BC036115.1 | - | 4 | 1482 | 5298999 |