Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005268208.1 | XP_005268265.1 | 15 | 1992 | ||
XM_005268209.3 | XP_005268266.1 | 15 | 1796 | |||
XM_017021765.1 | XP_016877254.1 | 15 | 1759 | |||
GenBank mRNA | AB082526.1 | - | 15 | 1928 | ||
AK128693.1 | - | 15 | 2396 | |||
AL832633.2 | - | 9 | 1617 | |||
AY048580.1 | - | 15 | 1822 | |||
AY080896.1 | - | 15 | 1834 | |||
BC093881.1 | - | 15 | 1903 | 7939726 | ||
BC112101.1 | - | 15 | 1903 | 8327569 |