Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152277.2 | NP_689490.2 | 2 | 407 | ||
XM_017010022.1 | XP_016865511.1 | 2 | 417 | |||
XM_017010023.1 | XP_016865512.1 | 2 | 341 | |||
GenBank mRNA | AK313274.1 | - | 2 | 373 | ||
BC019910.1 | - | 2 | 255 | 4402070 | ||
BU849695.1 | - | 2 | 422 |