Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080863.4 | NP_543139.4 | 2 | 591 | ||
GenBank mRNA | AF403034.1 | - | 1 | 206 | ||
AK054727.1 | - | 2 | 591 | |||
AK314595.1 | - | 2 | 583 | |||
AY557346.1 | - | 2 | 507 | |||
BC075088.2 | - | 2 | 540 | 30915388 | ||
BC118584.1 | - | 2 | 573 | 40084995 | ||
BC118585.1 | - | 2 | 573 | 40084992 |