Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113525.1 | NP_001106997.1 | 8, 9 | 1449 | ||
NM_152287.3 | NP_689500.2 | 8, 9 | 1439 | |||
NR_110122.1 | 8, 9 | 1439 | ||||
NR_110126.1 | 8, 9 | 1407 | ||||
NR_110128.1 | 8, 9 | 1330 | ||||
NR_110129.1 | 8, 9 | 1419 | ||||
XM_005256324.3 | XP_005256381.1 | 8, 9 | 1398 | |||
XM_005256328.3 | XP_005256385.1 | 6, 7 | 1079 | |||
XM_017023890.1 | XP_016879379.1 | 7, 8 | 1374 | |||
GenBank mRNA | AK026482.1 | - | 8, 9 | 1389 | ||
AK056405.1 | - | 8, 9 | 1390 | |||
AK096762.1 | - | 1, 2 | 2028 | |||
AK289801.1 | - | 8, 9 | 1298 | |||
AK295770.1 | - | 6, 7 | 1111 | |||
AK315793.1 | - | 5, 6 | 806 | |||
BC032781.1 | - | 7, 8 | 1108 | |||
BC105959.1 | - | 8, 9 | 1232 | 40036181 | ||
BC107603.1 | - | 8, 9 | 1264 | 40036180 | ||
BC110348.1 | - | 8, 9 | 1164 | 5196957 |