Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPLL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142650.1 | NP_001136122.1 | 5 | 733 | ||
NM_138394.3 | NP_612403.2 | 4 | 989 | |||
XM_005264639.3 | XP_005264696.2 | 4 | 986 | |||
XM_005264640.3 | XP_005264697.2 | 5 | 732 | |||
XM_011533165.2 | XP_011531467.1 | 4 | 986 | |||
XM_011533166.2 | XP_011531468.1 | 4 | 986 | |||
GenBank mRNA | AK000155.1 | - | 5 | 637 | ||
AK291462.1 | - | 4 | 854 | |||
AK292259.1 | - | 5 | 726 | |||
AL512692.1 | - | 3 | 383 | |||
AY236962.1 | - | 4 | 818 | |||
AY236963.2 | - | 5 | 578 | |||
BC008217.1 | - | 5 | 626 | 3449607 | ||
BC017480.1 | - | 4 | 835 | 3502860 | ||
KU178839.1 | - | 4 | 579 | |||
KU178840.1 | - | 4 | 579 | |||
KU178841.1 | - | 4 | 579 |