Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040272.5 | NP_001035362.3 | 4 | 428 | ||
NM_052866.4 | NP_443098.3 | 4 | 428 | |||
XM_011518063.2 | XP_011516365.1 | 6 | 566 | |||
XM_011518064.2 | XP_011516366.1 | 5 | 681 | |||
XM_017015310.1 | XP_016870799.1 | 5 | 7203 | |||
XM_017015311.1 | XP_016870800.1 | 6 | 566 | |||
XM_017015312.1 | XP_016870801.1 | 5 | 682 | |||
XM_017015313.1 | XP_016870802.1 | 5 | 329 | |||
XM_017015314.1 | XP_016870803.1 | 6 | 566 | |||
GenBank mRNA | AF176313.1 | - | 4 | 326 | ||
AK290667.1 | - | 4 | 414 | |||
AK291956.1 | - | 4 | 368 | |||
AY358327.1 | - | 4 | 408 | |||
BC030262.1 | - | 4 | 340 | 5201199 | ||
CD636819.1 | - | 2 | 216 | |||
CX758410.1 | - | 4 | 327 | |||
DA598045.1 | - | 4 | 403 |