Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127441.1 | NP_001120913.1 | 6 | 928 | ||
NM_001127442.1 | NP_001120914.1 | 4 | 817 | |||
NM_001318223.1 | NP_001305152.1 | 4 | 817 | |||
NM_080385.4 | NP_525124.3 | 5 | 875 | |||
XM_005250710.1 | XP_005250767.1 | 5 | 856 | |||
XM_005250712.1 | XP_005250769.1 | 4 | 770 | |||
XM_011516698.1 | XP_011515000.1 | 4 | 1493 | |||
XM_011516701.1 | XP_011515003.1 | 4 | 1410 | |||
XM_011516702.1 | XP_011515004.1 | 4 | 1396 | |||
XM_011516703.1 | XP_011515005.1 | 4 | 1389 | |||
XM_011516704.1 | XP_011515006.1 | 4 | 1507 | |||
XM_011516705.1 | XP_011515007.1 | 4 | 1507 | |||
GenBank mRNA | AF384667.1 | - | 3 | 308 | ||
AK093288.1 | - | 4 | 530 | |||
AK131191.1 | - | 4 | 753 | |||
AK315256.1 | - | 4 | 791 | |||
AY155229.1 | - | 5 | 828 | |||
AY155230.1 | - | 6 | 898 | |||
BC039362.1 | - | 6 | 902 | 5270617 | ||
BC042996.1 | - | 4 | 1444 | 5296768 |