Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032796.3 | NP_116185.2 | 8 | 1003 | ||
NR_033181.1 | 8 | 965 | ||||
GenBank mRNA | AF168955.1 | - | 8 | 955 | ||
AF305828.1 | - | 5 | 495 | |||
AF338728.1 | - | 8 | 955 | |||
AK027401.1 | - | 8 | 945 | |||
AK126173.1 | - | 3 | 1918 | |||
AK222856.1 | - | 8 | 937 | |||
AK295322.1 | - | 8 | 940 | |||
AK303783.1 | - | 8 | 919 | |||
AK314675.1 | - | 8 | 975 | |||
BC001468.1 | - | 8 | 934 | |||
BC014657.1 | - | 8 | 923 | 4640905 | ||
CR749849.1 | - | 8 | 989 |