Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPUSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324086.1 | NP_001311015.1 | 6 | 761 | ||
NM_001324410.1 | NP_001311339.1 | 6 | 662 | |||
NM_001324411.1 | NP_001311340.1 | 5 | 649 | |||
NM_001324412.1 | NP_001311341.1 | 5 | 563 | |||
NM_001324413.1 | NP_001311342.1 | 6 | 653 | |||
NM_001324414.1 | NP_001311343.1 | 6 | 709 | |||
NM_001324415.1 | NP_001311344.1 | 6 | 762 | |||
NM_058192.2 | NP_478072.1 | 6 | 752 | |||
GenBank mRNA | AL514840.3 | - | 6 | 612 | ||
AL520203.3 | - | 5 | 591 | |||
BC011783.2 | - | 5 | 617 | 3937184 | ||
BC012951.2 | - | 2 | 252 | |||
BC036279.2 | - | 2 | 252 | |||
BX358716.2 | - | 5 | 548 | |||
CX786721.1 | - | 6 | 712 |