Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015163.5 | NP_055978.4 | 5 | 1990 | ||
NM_052978.4 | NP_443210.1 | 5 | 1990 | |||
XM_005267301.3 | XP_005267358.1 | 6 | 2595 | |||
XM_005267302.3 | XP_005267359.1 | 6 | 2595 | |||
XM_006720018.3 | XP_006720081.1 | 6 | 2595 | |||
XM_011536389.2 | XP_011534691.1 | 6 | 2610 | |||
XM_017020943.1 | XP_016876432.1 | 6 | 2595 | |||
XM_017020944.1 | XP_016876433.1 | 6 | 2595 | |||
XM_017020945.1 | XP_016876434.1 | 6 | 2595 | |||
XM_017020946.1 | XP_016876435.1 | 6 | 2595 | |||
XM_017020947.1 | XP_016876436.1 | 6 | 2595 | |||
XM_017020948.1 | XP_016876437.1 | 6 | 2595 | |||
XM_017020949.1 | XP_016876438.1 | 6 | 2595 | |||
XM_017020950.1 | XP_016876439.1 | 6 | 2595 | |||
GenBank mRNA | AK055388.1 | - | 5 | 1615 | ||
BC013414.2 | - | 5 | 1362 | 3510374 | ||
BC063872.1 | - | 5 | 2347 | 6471552 | ||
D87458.2 | - | 5 | 1366 |