Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099272.1 | NP_001092742.1 | 4 | 1161 | ||
NM_001172418.1 | NP_001165889.1 | 3 | 946 | |||
NM_052893.1 | NP_443125.1 | 5 | 1361 | |||
NM_152733.2 | NP_689946.2 | 3 | 898 | |||
XM_011514279.2 | XP_011512581.1 | 6 | 1758 | |||
XM_011514280.2 | XP_011512582.1 | 4 | 1439 | |||
XM_011514281.2 | XP_011512583.1 | 4 | 1439 | |||
XM_011514282.2 | XP_011512584.1 | 4 | 1439 | |||
XM_017010227.1 | XP_016865716.1 | 5 | 1639 | |||
GenBank mRNA | AB067467.1 | - | 4 | 1027 | ||
AK057507.1 | - | 3 | 898 | |||
BC101354.2 | - | 3 | 819 | 40025568 | ||
BC101355.1 | - | 3 | 819 | 40025569 | ||
BC101356.2 | - | 3 | 819 | |||
BC101357.1 | - | 3 | 867 | 40025572 | ||
BX537534.1 | - | 4 | 894 |