Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281956.1 | NP_001268885.1 | 58 | 9309 | ||
NM_052896.4 | NP_443128.2 | 57 | 8729 | |||
XM_017000185.1 | XP_016855674.1 | 58 | 9228 | |||
XM_017000186.1 | XP_016855675.1 | 58 | 9228 | |||
XM_017000187.1 | XP_016855676.1 | 58 | 9057 | |||
XM_017000188.1 | XP_016855677.1 | 58 | 8966 | |||
XM_017000189.1 | XP_016855678.1 | 58 | 9228 | |||
XM_017000190.1 | XP_016855679.1 | 52 | 8493 | |||
XM_017000194.1 | XP_016855683.1 | 58 | 8966 | |||
GenBank mRNA | AB067471.1 | - | 7 | 1079 | ||
AB212622.1 | - | 58 | 9309 | |||
AY210418.1 | - | 57 | 8729 |