Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052900.2 | NP_443132.3 | 29 | 4850 | ||
NM_198123.1 | NP_937756.1 | 30 | 5162 | |||
NM_198124.1 | NP_937757.1 | 31 | 5054 | |||
XM_011516815.2 | XP_011515117.1 | 4 | 663 | |||
XM_011516816.2 | XP_011515118.1 | 29 | 5336 | |||
XM_017013008.1 | XP_016868497.1 | 29 | 5570 | |||
XM_017013009.1 | XP_016868498.1 | 30 | 5648 | |||
XM_017013010.1 | XP_016868499.1 | 28 | 5258 | |||
XM_017013011.1 | XP_016868500.1 | 28 | 5258 | |||
XM_017013012.1 | XP_016868501.1 | 28 | 4621 | |||
GenBank mRNA | AB067481.2 | - | 17 | 3023 | ||
AB114604.1 | - | 30 | 5162 | |||
AB114605.1 | - | 31 | 5054 | |||
AY210419.1 | - | 29 | 4721 |