Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052900.2 | NP_443132.3 | 43 | 6806 | ||
NM_198123.1 | NP_937756.1 | 44 | 7118 | |||
NM_198124.1 | NP_937757.1 | 45 | 7010 | |||
XM_011516815.2 | XP_011515117.1 | 18 | 2619 | |||
XM_011516816.2 | XP_011515118.1 | 43 | 7292 | |||
XM_017013008.1 | XP_016868497.1 | 43 | 7526 | |||
XM_017013009.1 | XP_016868498.1 | 42 | 7394 | |||
XM_017013010.1 | XP_016868499.1 | 42 | 7214 | |||
XM_017013011.1 | XP_016868500.1 | 40 | 7004 | |||
XM_017013012.1 | XP_016868501.1 | 42 | 6577 | |||
GenBank mRNA | AB067481.2 | - | 29 | 4769 | ||
AB114604.1 | - | 44 | 7118 | |||
AB114605.1 | - | 45 | 7010 | |||
AK126252.1 | - | 2 | 151 | |||
AY210419.1 | - | 43 | 6677 |