Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK11IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052902.3 | NP_443134.3 | 11 | 1068 | ||
XM_011510526.1 | XP_011508828.1 | 11 | 1048 | |||
XM_011510527.2 | XP_011508829.1 | 11 | 1048 | |||
XM_011510529.1 | XP_011508831.1 | 11 | 1048 | |||
GenBank mRNA | AB067485.1 | - | 9 | 746 | ||
AF450267.1 | - | 11 | 1022 | |||
AK091963.1 | - | 10 | 950 | |||
AK295609.1 | - | 10 | 1236 | |||
AK296632.1 | - | 11 | 1048 | |||
AK300611.1 | - | 10 | 955 | |||
BC014114.2 | - | 2 | 109 | |||
BC034051.1 | - | 11 | 1045 | 4819148 |