Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052911.2 | NP_443143.2 | 4 | 1303 | ||
XM_011525798.1 | XP_011524100.1 | 4 | 1433 | |||
XM_011525799.2 | XP_011524101.1 | 4 | 1566 | |||
GenBank mRNA | AB067498.1 | - | 4 | 1239 | ||
AK096746.1 | - | 4 | 1305 | |||
AL832041.1 | - | 4 | 1455 | |||
AL834200.1 | - | 11 | ||||
BC089426.1 | - | 1 | 723 | 3090307 |