Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LENG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052925.3 | NP_443157.1 | 6 | 868 | ||
XM_005278248.4 | XP_005278305.1 | 6 | 859 | |||
XM_005278249.4 | XP_005278306.1 | 6 | 1311 | |||
XM_005278250.4 | XP_005278307.1 | 5 | 730 | |||
XM_005278252.4 | XP_005278309.1 | 5 | 1200 | |||
XM_006722996.3 | XP_006723059.1 | 5 | 735 | |||
XM_006722997.3 | XP_006723060.1 | 4 | 624 | |||
XM_011526413.2 | XP_011524715.1 | 6 | 798 | |||
XM_011526414.2 | XP_011524716.1 | 5 | 748 | |||
XM_011526415.2 | XP_011524717.1 | 6 | 859 | |||
GenBank mRNA | AB067519.1 | - | 1991 | |||
BC028048.1 | - | 6 | 819 | 3907442 |