Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103150.1 | NP_001096620.1 | 2 | 881 | ||
NM_001103151.1 | NP_001096621.1 | 3 | 781 | |||
NM_001184924.1 | NP_001171853.1 | 4 | 866 | |||
NM_052926.2 | NP_443158.1 | 2 | 867 | |||
XM_017029252.1 | XP_016884741.1 | 4 | 1183 | |||
XM_017029253.1 | XP_016884742.1 | 3 | 845 | |||
GenBank mRNA | AB067521.1 | - | 2 | 864 | ||
AK295446.1 | - | 3 | 745 | |||
AK316104.1 | - | 3 | 750 | |||
AY121806.1 | - | 2 | 864 | |||
BC101111.1 | - | 2 | 725 | 40011824 | ||
BC101112.2 | - | 2 | 711 | 40011826 | ||
BC101113.1 | - | 2 | 711 | 40011827 |