Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022776.4 | NP_073613.2 | 8 | 1856 | ||
GenBank mRNA | AF392454.1 | - | 8 | 1427 | ||
AK023074.1 | - | 3 | 253 | |||
AK023226.1 | - | 3 | 286 | |||
AK292702.1 | - | 8 | 1208 | |||
BC010422.1 | - | 3 | 440 | |||
BC065213.1 | - | 8 | 1206 | 3916115 |