Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006615.2 | NP_001006616.1 | 7 | 782 | ||
NM_001012361.3 | NP_001012361.1 | 8 | 915 | |||
NM_145241.4 | NP_660284.1 | 8 | 912 | |||
NR_134677.1 | 8 | 844 | ||||
XM_011518194.2 | XP_011516496.1 | 7 | 826 | |||
GenBank mRNA | AK074891.1 | - | 7 | 696 | ||
AK093240.1 | - | 8 | 844 | |||
AK313843.1 | - | 8 | 854 | |||
AL833635.1 | - | 8 | 761 | |||
BC012352.1 | - | 8 | 844 | 4634445 | ||
BC022982.1 | - | 8 | 841 |