Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198075.3 | NP_932341.1 | 12 | 1626 | ||
XM_005252775.2 | XP_005252832.1 | 12 | 2450 | |||
XM_005252776.2 | XP_005252833.1 | 11 | 2273 | |||
XM_006718132.2 | XP_006718195.1 | 12 | 2525 | |||
XM_006718133.2 | XP_006718196.1 | 11 | 1333 | |||
XM_011519875.2 | XP_011518177.1 | 13 | 1865 | |||
XM_011519876.1 | XP_011518178.1 | 13 | 2521 | |||
XM_011519877.2 | XP_011518179.1 | 12 | 1602 | |||
XM_017017167.1 | XP_016872656.1 | 13 | 1940 | |||
XM_017017168.1 | XP_016872657.1 | 13 | 2179 | |||
GenBank mRNA | AK024495.1 | - | 9 | 3516 | ||
BC016439.1 | - | 10 | 1160 | 3447051 | ||
BC035937.1 | - | 12 | 1612 | 4816105 |