Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVI5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159944.2 | NP_001153416.1 | 6 | 842 | ||
NM_145245.4 | NP_660288.1 | 7 | 953 | |||
XM_005272458.4 | XP_005272515.1 | 7 | 947 | |||
XM_017026239.1 | XP_016881728.1 | 7 | 939 | |||
GenBank mRNA | AB449874.1 | - | 6 | 799 | ||
BC014111.2 | - | 7 | 871 | 4542709 |