Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138691.2 | NP_619636.2 | 17 | 1963 | ||
XM_017014256.1 | XP_016869745.1 | 14 | 1686 | |||
GenBank mRNA | AF417578.1 | - | 17 | 1963 | ||
AK098607.1 | - | 2 | 47 | |||
AK307461.1 | - | 15 | 1687 | |||
BC103955.1 | - | 12 | 1325 | |||
BC103956.1 | - | 12 | 1325 | |||
BC114380.1 | - | 12 | 1325 | |||
BC141860.1 | - | 12 | 1325 |