Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138691.2 | NP_619636.2 | 20 | 2454 | ||
XM_017014256.1 | XP_016869745.1 | 17 | 2177 | |||
GenBank mRNA | AF417578.1 | - | 20 | 2454 | ||
AK098607.1 | - | 7 | 812 | |||
AK307461.1 | - | 18 | 2178 | |||
BC103955.1 | - | 16 | 2020 | |||
BC103956.1 | - | 15 | 1816 | |||
BC114380.1 | - | 15 | 1816 | |||
BC141860.1 | - | 15 | 1816 |