Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135196.1 | NP_001128668.1 | 3 | 2093 | ||
XM_005269476.4 | XP_005269533.1 | 3 | 2158 | |||
XM_005269477.3 | XP_005269534.1 | 3 | 2078 | |||
XM_005269478.4 | XP_005269535.1 | 3 | 2146 | |||
XM_017015619.1 | XP_016871108.1 | 4 | 1868 | |||
GenBank mRNA | AK127811.1 | - | 3 | 2093 | ||
AL833265.1 | - | 3 | 2042 | |||
BC126413.1 | - | 1 | 1794 |