Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR5P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153444.1 | NP_703145.1 | 1 | 423 | ||
GenBank mRNA | AF173006.1 | - | 1 | 423 | ||
AF220494.1 | - | 1 | 84 | |||
BC101710.1 | - | 1 | 454 | 8069216 | ||
BC113557.1 | - | 1 | 454 | 8322609 |