Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANO4 SIRNA Assays ›
Gene Symbol:
ANO4
Gene Name:
anoctamin 4
Gene Aliases:
TMEM16D
Chromosome Location:
Chr.12: 100717457 - 101128641 on Build GRCh38
UniGene ID:
Hs.58785
Species:
Human
siRNA ID
HSS133556
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANO4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286615.1
NP_001273544.1
20, 21
2476
NM_001286616.1
NP_001273545.1
19, 20
2044
NM_178826.3
NP_849148.2
19, 20
2371
XM_011537911.2
XP_011536213.1
22, 23
2471
XM_011537912.2
XP_011536214.1
22, 23
2471
XM_011537913.2
XP_011536215.1
21, 22
2426
XM_011537914.2
XP_011536216.1
19, 20
1982
XM_011537915.2
XP_011536217.1
20, 21
2294
XM_011537916.2
XP_011536218.1
20, 21
2135
XM_017018808.1
XP_016874297.1
19, 20
1982
XM_017018809.1
XP_016874298.1
21, 22
2562
XM_017018810.1
XP_016874299.1
19, 20
2224
XM_017018811.1
XP_016874300.1
18, 19
2179
XM_017018812.1
XP_016874301.1
18, 19
4197
XM_017018813.1
XP_016874302.1
17, 18
2047
XM_017018814.1
XP_016874303.1
20, 21
4436
XM_017018815.1
XP_016874304.1
20, 21
4436
XM_017018816.1
XP_016874305.1
20, 21
2519
XM_017018817.1
XP_016874306.1
20, 21
4471
GenBank mRNA
AK091540.1
-
10, 11
992
AK091591.1
-
19, 20
2160
AK092596.1
-
7, 8
558
AK293810.1
-
20, 21
2114
AK316105.1
-
20, 21
2115
BC109308.2
-
19, 20
2029
40035759
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
chloride transport
lipid transport
ion transmembrane transport
calcium activated phosphatidylserine scrambling
calcium activated phosphatidylcholine scrambling
calcium activated galactosylceramide scrambling
chloride transmembrane transport
intracellular calcium activated chloride channel activity
phospholipid scramblase activity
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0