Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152335.2 | NP_689548.2 | 8, 9 | 912 | ||
XM_005254160.2 | XP_005254217.1 | 8, 9 | 835 | |||
XM_017021915.1 | XP_016877404.1 | 10, 11 | 1128 | |||
XM_017021916.1 | XP_016877405.1 | 10, 11 | 1074 | |||
XM_017021917.1 | XP_016877406.1 | 10, 11 | 1060 | |||
GenBank mRNA | AK095509.1 | - | 8, 9 | 804 | ||
AK308676.1 | - | 7, 8 | 730 | |||
BC104953.1 | - | 2, 3 | 239 | 8143956 | ||
BC104955.1 | - | 2, 3 | 239 | 8143958 | ||
DR007145.1 | - | 4, 5 | 382 |