Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242929.1 | NP_001229858.1 | 12 | 1644 | ||
NM_001308089.1 | NP_001295018.1 | 14 | 1834 | |||
NM_001324129.1 | NP_001311058.1 | 11 | 1472 | |||
NM_001324130.1 | NP_001311059.1 | 14 | 2037 | |||
NM_133450.3 | NP_597707.1 | 15 | 2036 | |||
NR_040252.1 | 14 | 2039 | ||||
XM_005255119.3 | XP_005255176.1 | 13 | 1736 | |||
XM_005255121.3 | XP_005255178.1 | 11 | 1434 | |||
XM_011522372.1 | XP_011520674.1 | 14 | 1966 | |||
XM_011522373.1 | XP_011520675.1 | 14 | 1848 | |||
XM_011522374.1 | XP_011520676.1 | 13 | 1582 | |||
XM_011522378.1 | XP_011520680.1 | 14 | 1704 | |||
XM_017022935.1 | XP_016878424.1 | 12 | 1644 | |||
XM_017022936.1 | XP_016878425.1 | 14 | 1957 | |||
GenBank mRNA | AB075857.1 | - | 12 | 2472 | ||
AK056907.1 | - | 4 | 1585 | |||
AK057329.1 | - | 11 | 2195 | |||
AK123496.1 | - | 15 | 2037 | |||
AK301683.1 | - | 13 | 1664 | |||
BC050387.1 | - | 14 | 1675 | 5751728 |