Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153209.3 | NP_694941.2 | 6, 7 | 726 | ||
XM_005257029.1 | XP_005257086.1 | 6, 7 | 726 | |||
XM_017024150.1 | XP_016879639.1 | 6, 7 | 726 | |||
XM_017024151.1 | XP_016879640.1 | 6, 7 | 726 | |||
XM_017024155.1 | XP_016879644.1 | 6, 7 | 726 | |||
XM_017024156.1 | XP_016879645.1 | 6, 7 | 726 | |||
GenBank mRNA | AK094619.1 | - | 6, 7 | 681 | ||
BC093631.1 | - | 6, 7 | 612 | 7939476 | ||
BC104852.1 | - | 6, 7 | 612 | 8143855 | ||
BC110989.1 | - | 6, 7 | 690 |