Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173479.3 | NP_775750.3 | 11 | 1394 | ||
NR_135565.1 | 12 | 1509 | ||||
XM_011526449.2 | XP_011524751.1 | 11 | 2747 | |||
GenBank mRNA | AI982942.1 | - | 1 | 328 | ||
AK131444.1 | - | 11 | 1753 | |||
BC031227.1 | - | 12 | 1507 |