Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323906.1 | NP_001310835.1 | 8, 9 | 1414 | ||
NM_001323907.1 | NP_001310836.1 | 7, 8 | 1180 | |||
NM_001323908.1 | NP_001310837.1 | 8, 9 | 1446 | |||
NM_001323909.1 | NP_001310838.1 | 7, 8 | 1300 | |||
NM_001323910.1 | NP_001310839.1 | 9, 10 | 1462 | |||
NM_001323911.1 | NP_001310840.1 | 7, 8 | 1300 | |||
NM_001323912.1 | NP_001310841.1 | 7, 8 | 1251 | |||
NM_001323913.1 | NP_001310842.1 | 7, 8 | 1246 | |||
NM_144618.2 | NP_653219.1 | 8, 9 | 1388 | |||
XM_017000246.1 | XP_016855735.1 | 9, 10 | 1892 | |||
XM_017000247.1 | XP_016855736.1 | 10, 11 | 1979 | |||
XM_017000248.1 | XP_016855737.1 | 8, 9 | 1288 | |||
XM_017000249.1 | XP_016855738.1 | 9, 10 | 1348 | |||
XM_017000250.1 | XP_016855739.1 | 7, 8 | 991 | |||
XM_017000251.1 | XP_016855740.1 | 7, 8 | 1174 | |||
XM_017000252.1 | XP_016855741.1 | 8, 9 | 1134 | |||
XM_017000253.1 | XP_016855742.1 | 7, 8 | 980 | |||
XM_017000254.1 | XP_016855743.1 | 6, 7 | 988 | |||
GenBank mRNA | AK092230.1 | - | 8, 9 | 1228 | ||
AK301882.1 | - | 8, 9 | 1019 | |||
AK313606.1 | - | 8, 9 | 1201 | |||
BC027033.1 | - | 8, 9 | 1388 | 5139830 |