Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC44A3 SIRNA Assays ›
Gene Symbol:
SLC44A3
Gene Name:
solute carrier family 44 member 3
Gene Aliases:
CTL3
Chromosome Location:
Chr.1: 94820342 - 94895247 on Build GRCh38
UniGene ID:
Hs.483423
Species:
Human
siRNA ID
HSS133971
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC44A3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114106.2
NP_001107578.1
8, 9
1003
NM_001258340.1
NP_001245269.1
8, 9
1003
NM_001258341.1
NP_001245270.1
8, 9
907
NM_001258342.1
NP_001245271.1
7, 8
895
NM_001258343.1
NP_001245272.1
7, 8
799
NM_001301079.1
NP_001288008.1
7, 8
842
NM_152369.4
NP_689582.2
7, 8
938
XM_005270440.2
XP_005270497.1
8, 9
898
XM_005270441.2
XP_005270498.1
8, 9
940
XM_005270442.1
XP_005270499.1
8, 9
994
XM_011540632.2
XP_011538934.1
7, 8
867
XM_011540633.2
XP_011538935.1
7, 8
705
XM_011540635.2
XP_011538937.1
8, 9
994
XM_011540636.2
XP_011538938.1
8, 9
898
XM_017000262.1
XP_016855751.1
7, 8
880
XM_017000263.1
XP_016855752.1
8, 9
940
XM_017000264.1
XP_016855753.1
8, 9
994
XM_017000265.1
XP_016855754.1
8, 9
898
XM_017000266.1
XP_016855755.1
8, 9
994
GenBank mRNA
AK300193.1
-
7, 8
736
AK301288.1
-
7, 8
744
AK302381.1
-
5, 6
587
AK303906.1
-
7, 8
874
AK313574.1
-
8, 9
891
AK316123.1
-
9, 10
848
AY358659.1
-
4, 5
457
BC033858.1
-
8, 9
966
BC053877.1
-
7, 8
861
5760394
BX648225.1
-
7, 8
867
Back To Top
More Information
Panther Classification:
Molecular Function -
transporter
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidylcholine biosynthetic process
choline transport
transmembrane transport
protein binding
choline transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0