Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080868.2 | NP_543144.1 | 2 | 578 | ||
XM_006710345.3 | XP_006710408.1 | 3 | 410 | |||
XM_006710346.3 | XP_006710409.1 | 3 | 337 | |||
GenBank mRNA | AF403035.1 | - | 2 | 438 | ||
AF429968.1 | - | 2 | 555 | |||
AK098606.1 | - | 2 | 578 | |||
BC036573.2 | - | 2 | 556 | 5297837 |